Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone oil

Catalog No. 378321250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-832-1250ML 250 mL
37-832-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-832-1250ML Supplier Sigma Aldrich Supplier No. 378321250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is a low viscosity, liquid polymerized, siloxane with organic side chains. Silicone oil exhibits antimicrobial activity against endopthalmitis causing microorganisms under in vitro conditions.Silicone oil has been used as an intraocular liquid in the treatment of retinal detachments.It may also be used as a two phase partitioning bioreactor to treat gas effluents.

Specifications

Boiling Point >140°C/0.002, mmHg
CAS 63148-62-9
Quantity 250 mL
Linear Formula [-Si(CH3)2 O-]n
MDL Number MFCD00132673
Synonym PDMS; Polydimethylsiloxane
Refractive Index n20/D 1.403 (literature)
Density 0.93 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.