Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone oil

Catalog No. 378380250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-838-0250ML 250 mL
37-838-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-838-0250ML Supplier Sigma Aldrich Supplier No. 378380250ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 500 cSt that is dependent on the chain length distribution. Its refractive index is ∼ 1.402 and dielectric strength is ∼ 14 kV/mm. It's surface tension tends to increase with an increase in the viscosity.Silicone oil can be used for a variety of application such as: heat transferring medium in chemical and petrochemical industries, a dielectric coolant, a lubricant and antiflatulent agent, protective coatings for construction materials, a cosmetic additive.

Specifications

CAS 63148-62-9
Density 0.97 g/mL (at 25°C)
Boiling Point >140°C (0.002 mmHg)
Linear Formula [-Si(CH3)2 O-]n
Refractive Index n20/D 1.403 (literature)
MDL Number MFCD00132673
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.