Learn More
TA Instruments Silicone Oil Vial, Supplied with DSC Thermal Conductivity Kit, Improves Thermal Contact Between Sample and DSC Sensor, Compatible with DSC Thermal Conductivity Kit
Catalog No. 5000580680 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Vial Silicone Oil is supplied with the DSC Thermal Conductivity Kit, part number 915064.901. It improves the thermal contact between the sample being measured and the DSC sensor.
Catalog No. 50-005-80680 Supplier TA Instruments Supplier No. 915068.901
Waters TA Instruments Vial Silicone Oil is supplied with the DSC Thermal Conductivity Kit, part number 915064.901. It improves the thermal contact between the sample being measured and the DSC sensor.