Learn More
Cole-Parmer Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 9.5, Pink; 5/Bag
Catalog No. NC0300539 Shop All Cole-Parmer Products
Encompass_Preferred
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers. Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*
Catalog No. NC0300539 Supplier Cole-Parmer Supplier No. 0629816
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers. Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*