Learn More
Andwin Scientific Hirschmann Silicone Adapter, 0.1ml to 200ml
SDP
Catalog No. NC0883886
Encompass_Preferred
The Hirschmann Silicone Adapter is an accessory for the Pipetus Pipette Filler, which is a product that provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL. The adapter is sold separately from the pipetus pipette filler.
Catalog No. NC0883886 Supplier Andwin Scientific Supplier No. 9903201
Add to Cart
Add to Cart
The Hirschmann Silicone Adapter is an accessory for the Pipetus Pipette Filler, which is a product that provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL. The adapter is sold separately from the pipetus pipette filler.