Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, Platinum-Cured Silicone, 1.42 mm ID; 50 ft
Catalog No. NC0224904
Encompass
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.
Catalog No. NC0224904 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9559034
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.