Learn More
Helix Medical Inc SILICONE PUMP TUBING 50 FT
Catalog No. NC0758537
Encompass_Preferred
0.188 x 0.438" Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.
Catalog No. NC0758537 Supplier Helix Medical Inc Supplier No. 60825112
0.188 x 0.438" Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.