Learn More
SAI Infusion Technologies Rat Femoral Vein Catheter - 76cm, for Rats 150g+
SDP
Catalog No. NC0704548
Encompass
Femoral Vein Catheter for Rats 125g or larger. 14" Silicone catheter with a 2" tip and a suture bead to anchor the catheter in place. Sterile. Access witn a 25G blunt needle. Product is for laboratory animal research only.
Catalog No. NC0704548 Supplier SAI Infusion Technologies Supplier No. RFV36
Femoral Vein Catheter for Rats 125g or larger. 14" Silicone catheter with a 2" tip and a suture bead to anchor the catheter in place. Sterile. Access witn a 25G blunt needle. Product is for laboratory animal research only.