Learn More
Electron Microscopy Sciences Cold Embedding Silicone Mold for Acylic Resin 25 mm
SDP
Catalog No. 50997436
Encompass_Preferred
This mold is for acrylic resins only and the best mounting cup for all of our acrylic resins. The flexible silicone rubber makes it very easy to remove the mount from the mounting cup.
Diameter: 25 mm
Catalog No. 50-997-436 Supplier Electron Microscopy Sciences Supplier No. 5101225
This mold is for acrylic resins only and the best mounting cup for all of our acrylic resins. The flexible silicone rubber makes it very easy to remove the mount from the mounting cup.
Diameter: 25 mm