Learn More
Electron Microscopy Sciences Cold Embedding Silicone Mold for Acylic Resin 32 mm
SDP
Catalog No. 501929677
Encompass_Preferred
This mold is for acrylic resins only and the best mounting cup for all of our acrylic resins. The flexible silicone rubber makes it very easy to remove the mount from the mounting cup.
Diameter: 32 mm
Catalog No. 50-192-9677 Supplier Electron Microscopy Sciences Supplier No. 5101232
This mold is for acrylic resins only and the best mounting cup for all of our acrylic resins. The flexible silicone rubber makes it very easy to remove the mount from the mounting cup.
Diameter: 32 mm