Learn More
Analytical Sales & Services Silicone Rubber Alignment Gaskets
Catalog No. 5000805125
Encompass
Silicone Rubber Alignment Gaskets\nfor use with Flow Electrolysis Cells\n\nColor: orange\nThickness: 1/8”\n\nQty/Pkg: 10
Catalog No. 50-008-05125 Supplier Analytical Sales & Services Supplier No. F1088504
Silicone Rubber Alignment Gaskets\nfor use with Flow Electrolysis Cells\n\nColor: orange\nThickness: 1/8”\n\nQty/Pkg: 10