Learn More
BioSpec Products SILICONE RUBBER CAP, 100/PK
SDP
Catalog No. NC0465419
Encompass_Preferred
These silicone rubber caps were specifically designed for use with our 2 ml stainless steel microvials. Use when beadbeating or grinding with steel beads
Catalog No. NC0465419 Supplier BioSpec Products Supplier No. 2008
These silicone rubber caps were specifically designed for use with our 2 ml stainless steel microvials. Use when beadbeating or grinding with steel beads