Learn More
Chemglass Life Sciences Silicone Rubber Heater, 3 Wide x 24.18 Long for 238mm OD Vessels, Boot Latch Closure, 375 Watt, 110VAC, 6ft Cord with IEC Two Prong Plug
SDP
Catalog No. 5000083283
Encompass
Silicone rubber heater with boot latch, lace-style closure for 4 and 6 neck VSA (vertical side arm) bioreactors. Heaters are 375 watts and include a 6ft cord with an IEC two prong plug. Vessel OD - 238mm (9.37in) Approx. Circumference - 747.7mm (29.4in)For Use with 3L, 6L, & 8L 4 and 6 Neck VSA Vessels
Catalog No. 50-000-83283 Supplier Chemglass Life Sciences Supplier No. CLS1408238
Silicone rubber heater with boot latch, lace-style closure for 4 and 6 neck VSA (vertical side arm) bioreactors. Heaters are 375 watts and include a 6ft cord with an IEC two prong plug. Vessel OD - 238mm (9.37in) Approx. Circumference - 747.7mm (29.4in)For Use with 3L, 6L, & 8L 4 and 6 Neck VSA Vessels