Learn More
Chemglass Life Sciences Silicone Rubber Heater, 5 Wide x 45.56 Long for 411mm OD Vessels, Boot Latch Closure, 375 Watt, 110VAC, 6ft Cord with IEC Two Prong Plug
SDP
Catalog No. 5000083264
Encompass
Silicone rubber heater with boot latch, lace-style closure for 4 and 6 neck VSA (vertical side arm) bioreactors. Heaters are 375 watts and include a 6ft cord with an IEC two prong plug. Vessel OD - 411mm (16.2in) Approx. Circumference - 1921.2mm (50.8in)For Use with 36L 4 and 6 Neck VSA Vessels
Catalog No. 50-000-83264 Supplier Chemglass Life Sciences Supplier No. CLS1408411
Silicone rubber heater with boot latch, lace-style closure for 4 and 6 neck VSA (vertical side arm) bioreactors. Heaters are 375 watts and include a 6ft cord with an IEC two prong plug. Vessel OD - 411mm (16.2in) Approx. Circumference - 1921.2mm (50.8in)For Use with 36L 4 and 6 Neck VSA Vessels