Learn More
Chemglass Life Sciences Silicone sealing mat, sterile - 24-well (square wells), nested (10/pack) - for 24-well microplates
SDP
Catalog No. 502733435
Encompass_Preferred
Chemglass silicone sealing mat is a sterile, research-grade silicone mat designed to seal 24-well microplates with square wells for storage and assay applications. It minimizes evaporation, is autoclavable, and can be reused following appropriate cleaning protocols.
  • Designed for 24-well plates with square wells.
  • Made from research-grade silicone to minimize evaporation.
  • Sterile; supplied nested in packs (10 per pack, 50 per case).
  • Autoclavable and reusable following a bleach wash and ethanol rinse protocol.
  • Helps maintain sample integrity across a wide temperature range.
Catalog No. 50-273-3435 Supplier Chemglass Life Sciences Supplier No. CGN3596003
Chemglass silicone sealing mat is a sterile, research-grade silicone mat designed to seal 24-well microplates with square wells for storage and assay applications. It minimizes evaporation, is autoclavable, and can be reused following appropriate cleaning protocols.
  • Designed for 24-well plates with square wells.
  • Made from research-grade silicone to minimize evaporation.
  • Sterile; supplied nested in packs (10 per pack, 50 per case).
  • Autoclavable and reusable following a bleach wash and ethanol rinse protocol.
  • Helps maintain sample integrity across a wide temperature range.