Learn More
Chemglass Life Sciences Silicone Sealing Mat for 24-Well Deep-Well Plates, Square Wells - Non-Sterile, 10 per Pack
SDP
Catalog No. 502733431
Encompass_Preferred
Silicone sealing mat designed to provide an airtight, reusable seal for 24-well square-well plates used in storage and assay workflows. It reduces evaporation and contamination while allowing easy access to individual wells.
  • Made from silicone.
  • Designed for 24-well plates with square wells.
  • Non-sterile.
  • Pack size: 10 per pack, 50 per case.
  • Reusable and reduces sample evaporation and cross-contamination.
Catalog No. 50-273-3431 Supplier Chemglass Life Sciences Supplier No. CGN3596013
Silicone sealing mat designed to provide an airtight, reusable seal for 24-well square-well plates used in storage and assay workflows. It reduces evaporation and contamination while allowing easy access to individual wells.
  • Made from silicone.
  • Designed for 24-well plates with square wells.
  • Non-sterile.
  • Pack size: 10 per pack, 50 per case.
  • Reusable and reduces sample evaporation and cross-contamination.