Learn More
Chemglass Life Sciences Chemglass Silicone Sealing Mat, 384-Well, Square Well, Sterile
SDP
Catalog No. 502733434
Encompass_Preferred
Sterile silicone sealing mat designed to provide a tight seal for 384-well, square-well microplates to reduce evaporation during storage and assay applications. Compression-molded research-grade silicone offers flexibility and chemical compatibility; mats are packaged 10 per pack and 50 per case.
  • Sterile, research-grade silicone provides a tight seal.
  • Packaged 10 per pack and 50 per case.
  • Compatible with 384-well, square-well microplates.
  • Minimizes evaporation across a wide temperature range.
  • Autoclavable and reusable after a bleach wash and ethanol rinse.
  • Suitable for storage and assay applications requiring leak-resistant sealing.
Catalog No. 50-273-3434 Supplier Chemglass Life Sciences Supplier No. CGN3596004
Sterile silicone sealing mat designed to provide a tight seal for 384-well, square-well microplates to reduce evaporation during storage and assay applications. Compression-molded research-grade silicone offers flexibility and chemical compatibility; mats are packaged 10 per pack and 50 per case.
  • Sterile, research-grade silicone provides a tight seal.
  • Packaged 10 per pack and 50 per case.
  • Compatible with 384-well, square-well microplates.
  • Minimizes evaporation across a wide temperature range.
  • Autoclavable and reusable after a bleach wash and ethanol rinse.
  • Suitable for storage and assay applications requiring leak-resistant sealing.