Learn More
Chemglass Life Sciences Nest Silicone Sealing Mat - 384-Well Square Wells, Non-Sterile
SDP
Catalog No. 502733430
Encompass_Preferred
Silicone sealing mat designed to provide an airtight, liquid-tight seal for 384-well square-well microplates. Made from research-grade silicone, the mat reduces evaporation and contamination during storage and assays; it is non-sterile and can be autoclaved and reused after appropriate decontamination.
  • Designed for 384-well square-well microplates.
  • Made from research-grade silicone.
  • Non-sterile; intended for laboratory use.
  • Reusable after bleach wash and ethanol rinse; autoclaveable.
  • Packaged 10 mats per pack, 50 packs per case.
  • Provides an airtight and liquid-tight seal to minimize evaporation.
Catalog No. 50-273-3430 Supplier Chemglass Life Sciences Supplier No. CGN3596014
Silicone sealing mat designed to provide an airtight, liquid-tight seal for 384-well square-well microplates. Made from research-grade silicone, the mat reduces evaporation and contamination during storage and assays; it is non-sterile and can be autoclaved and reused after appropriate decontamination.
  • Designed for 384-well square-well microplates.
  • Made from research-grade silicone.
  • Non-sterile; intended for laboratory use.
  • Reusable after bleach wash and ethanol rinse; autoclaveable.
  • Packaged 10 mats per pack, 50 packs per case.
  • Provides an airtight and liquid-tight seal to minimize evaporation.