Learn More
Chemglass Life Sciences Silicone Sealing Mat for 96-Well Round Plates, Sterile, 10 per Pack
SDP
Catalog No. 502733428
Encompass_Preferred
Sterile silicone sealing mat designed to seal 96-well round microplates and minimize evaporation during storage and thermal cycling. Made from research-grade silicone and supplied sterile in laboratory pack sizes.
  • Seals 96-well round plates to minimize evaporation and cross-contamination.
  • Each well accommodates approximately 2.0 mL.
  • Supplied sterile, 10 per pack and 50 per case.
  • Made from research-grade silicone resistant to common laboratory conditions.
  • Can be autoclaved and reused following a bleach wash and ethanol rinse protocol.
  • Dimensions not specified by manufacturer.
Catalog No. 50-273-3428 Supplier Chemglass Life Sciences Supplier No. CGN3596022
Add to Cart
Add to Cart
Sterile silicone sealing mat designed to seal 96-well round microplates and minimize evaporation during storage and thermal cycling. Made from research-grade silicone and supplied sterile in laboratory pack sizes.
  • Seals 96-well round plates to minimize evaporation and cross-contamination.
  • Each well accommodates approximately 2.0 mL.
  • Supplied sterile, 10 per pack and 50 per case.
  • Made from research-grade silicone resistant to common laboratory conditions.
  • Can be autoclaved and reused following a bleach wash and ethanol rinse protocol.
  • Dimensions not specified by manufacturer.