Learn More
Chemglass Life Sciences Chemglass Silicone Sealing Mat, 96-Well (Round Wells), Pre-Slit, Sterile
SDP
Catalog No. 502733429
Encompass_Preferred
Sterile silicone sealing mat designed to seal 96-well microplates with round wells. The pre-slit, pierceable silicone reduces evaporation and contamination during storage, incubation, and thermal cycling, while allowing easy access to individual wells.
  • Sterile, pre-slit silicone for easy pipette access
  • Compatible with 96-well round-well microplates
  • Reduces evaporation and cross-contamination
  • 1.0 mL nominal well volume
  • Packaged 10 per pack, 50 per case
Catalog No. 50-273-3429 Supplier Chemglass Life Sciences Supplier No. CGN3596021
Sterile silicone sealing mat designed to seal 96-well microplates with round wells. The pre-slit, pierceable silicone reduces evaporation and contamination during storage, incubation, and thermal cycling, while allowing easy access to individual wells.
  • Sterile, pre-slit silicone for easy pipette access
  • Compatible with 96-well round-well microplates
  • Reduces evaporation and cross-contamination
  • 1.0 mL nominal well volume
  • Packaged 10 per pack, 50 per case