Learn More
Chemglass Life Sciences Chemglass silicone sealing mat for 96-well square-well microplates, non-sterile, puncturable (CGN-3596-012)
SDP
Catalog No. 502733432
Encompass_Preferred
Chemglass silicone sealing mat designed to seal 96-well square-well microplates and minimize evaporation during storage and processing. Made from research-grade silicone, the mat is puncturable for sample access and can be autoclaved and reused after appropriate cleaning.
  • 96-well, square well format
  • Puncturable to allow sample access
  • Non-sterile; supplied 10 per pack, 50 per case
  • Made from research-grade silicone
  • Autoclavable and reusable after bleach wash and ethanol rinse
Catalog No. 50-273-3432 Supplier Chemglass Life Sciences Supplier No. CGN3596012
Chemglass silicone sealing mat designed to seal 96-well square-well microplates and minimize evaporation during storage and processing. Made from research-grade silicone, the mat is puncturable for sample access and can be autoclaved and reused after appropriate cleaning.
  • 96-well, square well format
  • Puncturable to allow sample access
  • Non-sterile; supplied 10 per pack, 50 per case
  • Made from research-grade silicone
  • Autoclavable and reusable after bleach wash and ethanol rinse