Learn More
Chemglass Life Sciences Silicone Sealing Mat for 96-Well Plates, Square Well, Non-Sterile
SDP
Catalog No. 502733433
Encompass_Preferred
Silicone sealing mat designed to seal 96-well square-well microplates and minimize evaporation during storage and processing. Made from research-grade silicone for durability and can be reused after appropriate cleaning; compatible with autoclaving when cleaned according to recommended protocols.
  • Compatible with 96-well square-well plates.
  • Made from research-grade silicone for durability.
  • Non-sterile; recommended cleaning with bleach and ethanol before reuse.
  • Autoclavable when cleaned according to protocol.
  • Packaged 10 per pack, 50 per case.
  • Reduces evaporation and provides a tight seal for storage and handling.
Catalog No. 50-273-3433 Supplier Chemglass Life Sciences Supplier No. CGN3596011
Silicone sealing mat designed to seal 96-well square-well microplates and minimize evaporation during storage and processing. Made from research-grade silicone for durability and can be reused after appropriate cleaning; compatible with autoclaving when cleaned according to recommended protocols.
  • Compatible with 96-well square-well plates.
  • Made from research-grade silicone for durability.
  • Non-sterile; recommended cleaning with bleach and ethanol before reuse.
  • Autoclavable when cleaned according to protocol.
  • Packaged 10 per pack, 50 per case.
  • Reduces evaporation and provides a tight seal for storage and handling.