Learn More
Chemglass Life Sciences Chemglass Silicone Sealing Mat for 96-Well Square Microplates, Sterile, Pierceable (CGN-3596-002)
SDP
Catalog No. 502733436
Encompass_Preferred
The Chemglass silicone sealing mat provides a sterile, pierceable seal for 96-well square microplates to help prevent evaporation and contamination during storage and assay workflows.
  • Provides a sterile, pierceable seal for 96-well square well microplates.
  • Made from compression-molded silicone.
  • Prevents evaporation and contamination during storage and thermal cycling.
  • Suitable for use with 96-well microplates in storage and assay workflows.
  • Sold in 10 per pack, 50 per case quantities.
Catalog No. 50-273-3436 Supplier Chemglass Life Sciences Supplier No. CGN3596002
The Chemglass silicone sealing mat provides a sterile, pierceable seal for 96-well square microplates to help prevent evaporation and contamination during storage and assay workflows.
  • Provides a sterile, pierceable seal for 96-well square well microplates.
  • Made from compression-molded silicone.
  • Prevents evaporation and contamination during storage and thermal cycling.
  • Suitable for use with 96-well microplates in storage and assay workflows.
  • Sold in 10 per pack, 50 per case quantities.