Learn More
Chemglass Life Sciences Silicone Sealing Mat for 96-Well Microplates, Square Wells, Sterile
SDP
Catalog No. 502733437
Encompass_Preferred
Sterile silicone sealing mat designed to seal 96-well microplates with square wells, minimizing evaporation and protecting samples during storage and assay workflows. Sold in packs of 10 and cases of 50, it is autoclavable and can be reused following recommended cleaning protocols.
  • Sterile, research-grade silicone construction
  • Fits 96-well microplates with square wells
  • Pack size: 10 per pack; 50 per case
  • Autoclavable for laboratory sterilization
  • Reusable after bleach wash and ethanol rinse
  • Reduces evaporation during storage and assays
Catalog No. 50-273-3437 Supplier Chemglass Life Sciences Supplier No. CGN3596001
Sterile silicone sealing mat designed to seal 96-well microplates with square wells, minimizing evaporation and protecting samples during storage and assay workflows. Sold in packs of 10 and cases of 50, it is autoclavable and can be reused following recommended cleaning protocols.
  • Sterile, research-grade silicone construction
  • Fits 96-well microplates with square wells
  • Pack size: 10 per pack; 50 per case
  • Autoclavable for laboratory sterilization
  • Reusable after bleach wash and ethanol rinse
  • Reduces evaporation during storage and assays