Learn More
Grainger Silicone Sheet Food Grade, 12" Width x 12" Length x 0.125" Thick, Translucent White
Catalog No. NC2414475
Encompass_Preferred
The Approved Vendor Silicone Sheet Food Grade is a 12" x 12" x 0.125" thick silicone sheet made of food grade silicone. It has a durometer hardness of 40A and can withstand temperatures from -100 to 500°F. The silicone sheet is flexible, stain resistant, and FDA compliant, making it suitable for use as a baking mat, jar opener, trivet, and more in commercial and residential kitchens. With proper care, the silicone can be cut, folded, or rolled.
Catalog No. NC2414475 Supplier Grainger Supplier No. 241VK0
The Approved Vendor Silicone Sheet Food Grade is a 12" x 12" x 0.125" thick silicone sheet made of food grade silicone. It has a durometer hardness of 40A and can withstand temperatures from -100 to 500°F. The silicone sheet is flexible, stain resistant, and FDA compliant, making it suitable for use as a baking mat, jar opener, trivet, and more in commercial and residential kitchens. With proper care, the silicone can be cut, folded, or rolled.