Learn More
LabStrong Corp SILICONE SLEEVE OUTER
Catalog No. NC0614516
Encompass_Preferred
LabStrong part number 562X1LS compares to Fi-Streem part number 562X1. The glass condenser is a replacement part for Fi-Streem Water Purification Stills. The glass condenser comes with one 562X77LS (Silicone Sleeve, Large, 50mm length), one 562X64LS (Silicone Sleeve, Small, 35mm length), and one 562X63LS (Silicone Sleeve, Large, 60mm length).
This item is not returnable. View return policy
Catalog No. NC0614516 Supplier LabStrong Corp Supplier No. 562X77LS
LabStrong part number 562X1LS compares to Fi-Streem part number 562X1. The glass condenser is a replacement part for Fi-Streem Water Purification Stills. The glass condenser comes with one 562X77LS (Silicone Sleeve, Large, 50mm length), one 562X64LS (Silicone Sleeve, Small, 35mm length), and one 562X63LS (Silicone Sleeve, Large, 60mm length).