Learn More
Crescent Chemical Co Inc Silicon Solution Serva for Siconizing Glass and Metal, 100mL
SDP
Catalog No. 50248140
Encompass_Preferred
Silicon Solution Serva for Siconizing Glass and Metal, 100mL, in isopropanol, Suitable for siliconizing UV quartz cuvettes. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-140 Supplier Crescent Chemical Co Inc Supplier No. SE35130.01
Silicon Solution Serva for Siconizing Glass and Metal, 100mL, in isopropanol, Suitable for siliconizing UV quartz cuvettes. Storage Conditions: +15 °C TO +30 °C