Learn More
Crescent Chemical Co Inc SILICONE SOLUTION SERVA FOR
SDP
Catalog No. 50248142
Encompass_Preferred
Silicone solution SERVA for siliconizing glass and metal in isopropanol Suitable for siliconizing UV quartz cuvettes. Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 35130.03
Catalog No. 50-248-142 Supplier Crescent Chemical Co Inc Supplier No. SE35130.03
Silicone solution SERVA for siliconizing glass and metal in isopropanol Suitable for siliconizing UV quartz cuvettes. Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 35130.03