Learn More
Bellco Glass, Inc. Silicone Sponge Closure 13-18mm
SDP
Catalog No. 501945238
Encompass_Preferred
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 13-18mm Culture Tubes And 18mm Screw ThreadsCase Quantity: 10
Catalog No. 50-194-5238 Supplier Bellco Glass, Inc. Supplier No. 200400002
Sealed in silicone sponge closure is ideal for aerobic cultures. Flanged to fit fit snugly over outside of the neck preventing passage of aerosols. Silicone sponge closures can withstand dry heat sterilization up to 180 degrees Celsius. Use mainly for flasks, to prevent any trouble due to tumbling of the flask.Specifications:Cap TypeAir Permeation: 200-300 (Cc/Min)For Use With: 13-18mm Culture Tubes And 18mm Screw ThreadsCase Quantity: 10