Learn More
Bellco Glass, Inc. Silicone Sponge Closure,32mm Tapered
SDP
Catalog No. 501944696
Encompass_Preferred
The silicone sponge closure is suitable for cultivation of aerobic microorganisms. Can withstand heat up to 350 degree Celsius, if it lasts only for a short time so that the flame sterilization can be performed. Can be used the same way as a cotton plug f
Catalog No. 50-194-4696 Supplier Bellco Glass, Inc. Supplier No. 192400032
The silicone sponge closure is suitable for cultivation of aerobic microorganisms. Can withstand heat up to 350 degree Celsius, if it lasts only for a short time so that the flame sterilization can be performed. Can be used the same way as a cotton plug f