Learn More
Sigma Aldrich Fine Chemicals Biosciences Silicone sponge closures s
Catalog No. 502905771
Encompass
Silicone rubber rings with sealed-in silicone sponge filters.These silicone sponge closures are ideal when working with aerobic cultures they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180C and are easy to clean and maintain (reusable).
Catalog No. 50-290-5771 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C092110EA
Silicone rubber rings with sealed-in silicone sponge filters.These silicone sponge closures are ideal when working with aerobic cultures they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180C and are easy to clean and maintain (reusable).