Learn More
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 2, Pink; 20/Bag
Catalog No. NC0747299 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 2, Pink; 20/Bag, Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C).
Catalog No. NC0747299 Supplier Cole-Parmer Supplier No. 0629806
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 2, Pink; 20/Bag, Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C).