Learn More
Accuflow Systems Saint-Gobain Versilic Stopper, 56 x 65 x 45 mm, Silicone
Supplier:  Accuflow Systems D1069823
Encompass_Preferred
Saint-Gobain Versilic silicone stopper engineered for laboratory bottle and vessel closures. Molded from high-purity silicone elastomer, the stopper provides a resilient, chemically resistant seal and a smooth finish suitable for repeated use. Sized for secure fitting, the stopper measures 56 x 65 x 45 mm (top x bottom x height).
  • Made from high-purity silicone elastomer for broad chemical compatibility.
  • Dimensions 56 x 65 x 45 mm (top x bottom x height) for secure fit.
  • Provides a flexible, leak-resistant seal for bottles and vessels.
  • Withstands a wide temperature range suitable for common laboratory processes.
  • Smooth molded finish resists contamination and is easy to clean.
Catalog No. NC2327745