Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, BioPharm Platinum-Cured Silicone, I/P 73; 100 ft
Catalog No. NC0135644
Encompass
Choose our BioPharm Silicone platinum-cured tubing for purity, very low extractables, and an ultra-smooth inner surface. Ideal for pharmaceutical and biotechnological applications, this tubing complies with USP class VI and 3A sanitary standards. The ultra-smooth inner surface minimizes particle entrapment and microscopic build-up during fluid transfer. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.
Catalog No. NC0135644 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9642373
Choose our BioPharm Silicone platinum-cured tubing for purity, very low extractables, and an ultra-smooth inner surface. Ideal for pharmaceutical and biotechnological applications, this tubing complies with USP class VI and 3A sanitary standards. The ultra-smooth inner surface minimizes particle entrapment and microscopic build-up during fluid transfer. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.