Learn More
Bergen Industrial Supply SILICONE TC GASKET .75IN 50PK
Catalog No. NC9787162
Encompass_Preferred
Gasket, Tube Size 3/4 In, Tri-Clamp Connection, Material of Construction Platinum Silicone, Temp. Range -40F to 450F, Standards FDA, CLASS VI. 50/pack.
Catalog No. NC9787162 Supplier Bergen Industrial Supply Supplier No. 42RXPX075
Gasket, Tube Size 3/4 In, Tri-Clamp Connection, Material of Construction Platinum Silicone, Temp. Range -40F to 450F, Standards FDA, CLASS VI. 50/pack.