Learn More
Perkin Elmer US LLC Silicone Torch O-Ring, 9.25 mm I.D. for Optima 3x00 XL/3x00 DV/3000 SCX/4300/5300/7300 V
Catalog No. 502113061
Encompass_Preferred
Two silicone 9.25 mm I.D. O-rings are included in the Complete O-ring Kit for Optima 4300V/5300V/7300V (N0770916).
Catalog No. 50-211-3061 Supplier Perkin Elmer US LLC Supplier No. 09902247
Two silicone 9.25 mm I.D. O-rings are included in the Complete O-ring Kit for Optima 4300V/5300V/7300V (N0770916).