Learn More
Med Vet International Silicone Tracheostomy Tube, 8.0mm Uncuffed, J0690K
Catalog No. 5000582165
Encompass_Preferred
8.0mm i.d. x 11.3mm o.d. Uncuffed Tube
Catalog No. 50-005-82165 Supplier Med Vet International Supplier No. JOR690K
8.0mm i.d. x 11.3mm o.d. Uncuffed Tube