Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Platinum-Cured Silicone, 1/8"" ID x 1/4"" OD; 25 ft
Catalog No. NC9118666
Encompass
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications. Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI; exceeds 3A Sanitary Standards. Tubing is manufactured according to GMP. Temperature range: -58 to 446¿F (-50 to 230¿C) Sterilize: Autoclave or gamma irradiation.
Catalog No. NC9118666 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9580205
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications. Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI; exceeds 3A Sanitary Standards. Tubing is manufactured according to GMP. Temperature range: -58 to 446¿F (-50 to 230¿C) Sterilize: Autoclave or gamma irradiation.