Learn More
Accuflow Systems SILICONE TUB. 2-STOP #14 8/PK
Catalog No. 501108875
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex two-stop pump tubing, silicone (platinum-cured), L/S 14. Pack of 8. For use with Masterflex multichannel pump heads Specifications: Product Type - Stopped Pump Tubing Tubing formulation -
Catalog No. 50-110-8875 Supplier Accuflow Systems Supplier No. 0642114
Manufactured by Cole Parmer and sold by Accuflow. Masterflex two-stop pump tubing, silicone (platinum-cured), L/S 14. Pack of 8. For use with Masterflex multichannel pump heads Specifications: Product Type - Stopped Pump Tubing Tubing formulation -