Learn More
Accuflow Systems SILICONE TUB. 2-STOP 2.79 6/PK
Catalog No. 501109017
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Microbore two-stop tube sets, silicone (platinum-cured); 2.79 mm ID. Pk of 6. Specifications: Product Type - Stopped Pump Tubing Tubing formulation - Silicone (platinum-cured) Pump Series - L/S Masterflex
Catalog No. 50-110-9017 Supplier Accuflow Systems Supplier No. 0642148
Manufactured by Cole Parmer and sold by Accuflow. Microbore two-stop tube sets, silicone (platinum-cured); 2.79 mm ID. Pk of 6. Specifications: Product Type - Stopped Pump Tubing Tubing formulation - Silicone (platinum-cured) Pump Series - L/S Masterflex