Learn More
Bellco Glass, Inc. Silicone TUBING,1/2ID 3/4" OD
SDP
Catalog No. 501946259
Encompass_Preferred
Silicone tubing 1/2"x ¾". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Size:½” x 10'
Catalog No. 50-194-6259 Supplier Bellco Glass, Inc. Supplier No. 780010100
Silicone tubing 1/2"x ¾". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Size:½” x 10'