Learn More
Bellco Glass, Inc. Silicone TUBING,1/4ID 50 FT
SDP
Catalog No. 501946258
Encompass_Preferred
Silicone tubing 50ft. Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Size:¼” x 50'
Catalog No. 50-194-6258 Supplier Bellco Glass, Inc. Supplier No. 780010050
Silicone tubing 50ft. Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Size:¼” x 50'