Learn More
Ibidi USA Silicone Tubing 1.6 mm ID
SDP
Catalog No. NC0571368
Encompass
1.6 mm ID biocompatible silicone tubing used for a range of flow applications. silicone, sterile, autoclavable, 5 meters
This item is not returnable. View return policy
Catalog No. NC0571368 Supplier Ibidi USA Supplier No. 10842
1.6 mm ID biocompatible silicone tubing used for a range of flow applications. silicone, sterile, autoclavable, 5 meters