Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S 2-Stop Precision Pump Tubing, Platinum-Cured Silicone, L/S 13; Pack of 8
Catalog No. NC9375438
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent For use with Masterflex multichannel pump heads.
Catalog No. NC9375438 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642113
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent For use with Masterflex multichannel pump heads.