Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S 2-Stop Precision Pump Tubing, Platinum-Cured Silicone, L/S 16; Pack of 8
Catalog No. NC9375439
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent For use with Masterflex multichannel pump heads.
Catalog No. NC9375439 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642116
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent For use with Masterflex multichannel pump heads.