Learn More
A-M Systems SILICONE TUBING, 20FT
SDP
Catalog No. NC2263020
Encompass_Preferred
Silicone Tubing is a flexible, translucent tubing which contains no additives or plasticizers, and is non-reactive to tissue. The tubing surface is non-wetting to ensure smooth flow.
This item is not returnable. View return policy
Catalog No. NC2263020 Supplier A-M Systems Supplier No. 806100
Silicone Tubing is a flexible, translucent tubing which contains no additives or plasticizers, and is non-reactive to tissue. The tubing surface is non-wetting to ensure smooth flow.