Learn More
Bellco Glass, Inc. Silicone Tubing 25', 1/8OD Packs of 25'
SDP
Catalog No. 501946261
Encompass_Preferred
Silicone Tubing 1/8". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubingod: 1/8 (3.2Mm)id: 1/16" (1.6Mm)wall: 1/32" (0.8Mm)max.Pressure @ 70°c: 15packed 25' roll. Size:1/8” x 25'
Catalog No. 50-194-6261 Supplier Bellco Glass, Inc. Supplier No. 780010300
Silicone Tubing 1/8". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubingod: 1/8 (3.2Mm)id: 1/16" (1.6Mm)wall: 1/32" (0.8Mm)max.Pressure @ 70°c: 15packed 25' roll. Size:1/8” x 25'