Learn More
Bellco Glass, Inc. Silicone Tubing 25', 3/32OD Pkg of 25'
SDP
Catalog No. 501946260
Encompass_Preferred
Silicone Tubing is specifically formulated for use with media foundin pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubing od: 3/32" (2.4Mm)id: 1/32" (0.8Mm)wall: 1/32" (0.8Mm)max.Pressure @ 70°c: 31packed 25' roll. Size:3/32” x 25'
Catalog No. 50-194-6260 Supplier Bellco Glass, Inc. Supplier No. 780010200
Silicone Tubing is specifically formulated for use with media foundin pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubing od: 3/32" (2.4Mm)id: 1/32" (0.8Mm)wall: 1/32" (0.8Mm)max.Pressure @ 70°c: 31packed 25' roll. Size:3/32” x 25'