Learn More
Helix Medical Inc SILICONE TUBING 25'
Catalog No. NC1250054
Encompass_Preferred
0.099 x 0.139" Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing. Peristaltic pump tubing undergoes a Special Stabilization Process (SSP), providing the additional benefits of superior strength, performance, and longer life.
Catalog No. NC1250054 Supplier Helix Medical Inc Supplier No. 6082563
0.099 x 0.139" Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing. Peristaltic pump tubing undergoes a Special Stabilization Process (SSP), providing the additional benefits of superior strength, performance, and longer life.