Learn More
Helix Medical Inc SILICONE TUBING 25'
Supplier:  Helix Medical Inc 6082570
Encompass_Preferred
0.118 x 0.158" latinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing. Tubing is packaged double-bagged in crush-resistant boxes.
Catalog No. NC1250058